4-Cyan-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Cyano-2-fluorophenylboronic Acid, 4-Cyano-2-fluorobenzeneboronic acid

BB-9007-1G BB-9007-250M
COBBBB-9007-1GEA 473 EUR
COBBBB-9007-1G COBBBB-9007-250M
4-Cyan-2-fluorphenylboronsäure
4-Cyan-2-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164,93 g/mol
MDL: MFCD07644472
CAS-Nummer: 1150114-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR