4-Cyan-3-fluorphenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Cyano-3-fluorobenzeneboronic acid

H27710.14 H27710.03
H27710.14EA 692 EUR
H27710.14 H27710.03
4-Cyan-3-fluorphenylboronsäure ≥97%
4-Cyan-3-fluorphenylboronsäure

Purity: 97% CAS No.: 843663-18-3 Molecular Formula: C7H5BFNO2 Formula Weight: 164.93 Melting Point: 300° MDL No.: MFCD03411549

Formel: C₇H₅BFNO₂
Molecular Weight: 164.93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03411549
CAS-Nummer: 843663-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR