4-Cyan-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Cyano-3-fluorobenzeneboronic acid

BB-3408-1G BB-3408-25G BB-3408-5G
COBBBB-3408-1GEA 63.9 EUR
COBBBB-3408-1G COBBBB-3408-25G COBBBB-3408-5G
4-Cyan-3-fluorphenylboronsäure
4-Cyan-3-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164,93 g/mol
MDL: MFCD03411549
CAS-Nummer: 843663-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR