3-Fluor-4-(methoxy(methyl)carbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluoro-4-[methoxy(methyl)carbamoyl]phenylboronic acid, (3-Fluoro-4-(methoxy(methyl)carbamoyl)phenyl)boronic acid

BB-3439-5G BB-3439-1G
COBBBB-3439-5GEA 0 EUR
COBBBB-3439-5G COBBBB-3439-1G
3-Fluor-4-(methoxy(methyl)carbamoyl)phenylboronsäure
3-Fluor-4-(methoxy(methyl)carbamoyl)phenylboronsäure
Formel: C₉H₁₁BFNO₄
MDL: MFCD08689477
CAS-Nummer: 913835-59-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR