4-Fluor-3-(methoxy(methyl)carbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-3-[(N-methoxy-N-methyl)carbamoyl]phenylboronic acid, (4-Fluoro-3-(methoxy(methyl)carbamoyl)phenyl)boronic acid, 4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid

BB-3369-5G BB-3369-1G
COBBBB-3369-5GEA 451 EUR
COBBBB-3369-5G COBBBB-3369-1G
4-Fluor-3-(methoxy(methyl)carbamoyl)phenylboronsäure
4-Fluor-3-(methoxy(methyl)carbamoyl)phenylboronsäure
Formel: C₉H₁₁BFNO₄
Molecular Weight: 227.00 g/mol
MDL: MFCD08235106
CAS-Nummer: 874290-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR