2-Fluor-5-(methoxy(methyl)carbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: N-Methoxy-N-methyl 3-borono-4-fluorobenzamide

BB-3353-5G BB-3353-1G
COBBBB-3353-5GEA 0 EUR
COBBBB-3353-5G COBBBB-3353-1G
2-Fluor-5-(methoxy(methyl)carbamoyl)phenylboronsäure
2-Fluor-5-(methoxy(methyl)carbamoyl)phenylboronsäure
Formel: C₉H₁₁BFNO₄
MDL: MFCD08436038
CAS-Nummer: 874289-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR