3-Fluor-4-(2-hydroxyethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluoro-4-(2-hydroxyethylcarbamoyl)benzeneboronic acid

BB-3317-5G BB-3317-1G
COBBBB-3317-5GEA 0 EUR
COBBBB-3317-5G COBBBB-3317-1G
3-Fluor-4-(2-hydroxyethylcarbamoyl)phenylboronsäure
3-Fluor-4-(2-hydroxyethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₁BFNO₄
Molecular Weight: 227.00 g/mol
MDL: MFCD08689535
CAS-Nummer: 874289-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR