4-(Cyclohexylcarbamoyl)-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Cyclohexylcarbamoyl)-3-fluorophenyl)boronic acid

BB-3307-5G BB-3307-1G
COBBBB-3307-5GEA 1130 EUR
COBBBB-3307-5G COBBBB-3307-1G
4-(Cyclohexylcarbamoyl)-3-fluorphenylboronsäure
4-(Cyclohexylcarbamoyl)-3-fluorphenylboronsäure
Formel: C₁₃H₁₇BFNO₃
Molecular Weight: 265.09 g/mol
MDL: MFCD08436036
CAS-Nummer: 874289-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR