4-(Cyclopropylcarbamoyl)-3-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-(Cyclopropylcarbamoyl)-3-fluorophenyl)boronic acid
BB-3316-1G
BB-3316-5G
COBBBB-3316-1GEA
312
EUR
COBBBB-3316-1G
COBBBB-3316-5G
4-(Cyclopropylcarbamoyl)-3-fluorphenylboronsäure
4-(Cyclopropylcarbamoyl)-3-fluorphenylboronsäure
Formel:
C₁₀H₁₁BFNO₃ Molecular Weight: 223,0100 g/mol |
MDL:
MFCD09972092 CAS-Nummer: 874289-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...