3-Fluor-4-(methoxycarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Fluoro-4-(methoxycarbamoyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbamoyl)benzeneboronic acid

BB-3438-5G BB-3438-1G
COBBBB-3438-5GEA 0 EUR
COBBBB-3438-5G COBBBB-3438-1G
3-Fluor-4-(methoxycarbamoyl)phenylboronsäure
3-Fluor-4-(methoxycarbamoyl)phenylboronsäure
Formel: C₈H₉BFNO₄
MDL: MFCD08436059
CAS-Nummer: 913835-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR