4-Fluor-3-(methoxycarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Fluoro-3-(methoxycarbamoyl)benzeneboronic acid
BB-3367-5G
BB-3367-1G
COBBBB-3367-5GEA
595
EUR
COBBBB-3367-5G
COBBBB-3367-1G
4-Fluor-3-(methoxycarbamoyl)phenylboronsäure
4-Fluor-3-(methoxycarbamoyl)phenylboronsäure
Formel:
C₈H₉BFNO₄ Molecular Weight: 212.97 g/mol |
MDL:
MFCD08235105 CAS-Nummer: 913835-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...