3-Fluor-4-(2-pyridylcarbamoyl)phenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid

H54122.MD H54122.03
H54122.MDEA 139 EUR
H54122.MD H54122.03
3-Fluor-4-(2-pyridylcarbamoyl)phenylboronsäure ≥97%
3-Fluor-4-(2-pyridylcarbamoyl)phenylboronsäure

Purity: 97% Molecular Formula: C12H10BFN2O3 Formula Weight: 260.03 MDL No.: MFCD20265181

Formel: C₁₂H₁₀BFN₂O₃
Molecular Weight: 260.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20265181
CAS-Nummer: 1449133-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR