N,N-Diethyl-2-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4515-25G CA-4515-5G CA-4515-1G
COBBCA-4515-25GEA 884 EUR
COBBCA-4515-25G COBBCA-4515-5G COBBCA-4515-1G
N,N-Diethyl-2-fluorbenzamid
N,N-Diethyl-2-fluorbenzamid
Formel: C₁₁H₁₄FNO
MDL: MFCD00460287
CAS-Nummer: 10345-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR