4-Fluor-N,N-dimethylbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: N,N-Dimethyl 4-fluorobenzamide

CA-4502-5G CA-4502-25G CA-4502-1G
COBBCA-4502-5GEA 293 EUR
COBBCA-4502-5G COBBCA-4502-25G COBBCA-4502-1G
4-Fluor-N,N-dimethylbenzamid
4-Fluor-N,N-dimethylbenzamid
Formel: C₉H₁₀FNO
Molecular Weight: 167.18 g/mol
MDL: MFCD01214152
CAS-Nummer: 24167-56-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR