3-Fluor-N-methylbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: N-Methyl 3-fluorobenzamide

CA-4662-5G CA-4662-25G CA-4662-1G
COBBCA-4662-5GEA 385 EUR
COBBCA-4662-5G COBBCA-4662-25G COBBCA-4662-1G
3-Fluor-N-methylbenzamid
3-Fluor-N-methylbenzamid
Formel: C₈H₈FNO
MDL: MFCD06250758
CAS-Nummer: 701-39-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR