4-Fluor-N-tert-butylbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: N-t-Butyl-4-fluorobenzamide

CA-4777-25G CA-4777-1G CA-4777-5G
COBBCA-4777-25GEA 595 EUR
COBBCA-4777-25G COBBCA-4777-1G COBBCA-4777-5G
4-Fluor-N-tert-butylbenzamid
4-Fluor-N-tert-butylbenzamid
Formel: C₁₁H₁₄FNO
Molecular Weight: 195,24 g/mol
MDL: MFCD00463095
CAS-Nummer: 49834-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR