4-Fluor-N-methylbenzamid
Lieferant: COMBI-BLOCKS
CA-4503-1G
CA-4503-5G
CA-4503-25G
COBBCA-4503-1GEA
74.2
EUR
COBBCA-4503-1G
COBBCA-4503-5G
COBBCA-4503-25G
4-Fluor-N-methylbenzamid
4-Fluor-N-methylbenzamid
Formel:
C₈H₈FNO Molecular Weight: 153,1536 g/mol |
MDL:
MFCD00032531 CAS-Nummer: 701-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...