4-Fluor-N-propylbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: N-Propyl 4-fluorobenzamide

CA-4749-1G CA-4749-25G CA-4749-5G
COBBCA-4749-1GEA 74.2 EUR
COBBCA-4749-1G COBBCA-4749-25G COBBCA-4749-5G
4-Fluor-N-propylbenzamid
4-Fluor-N-propylbenzamid
Formel: C₁₀H₁₂FNO
MDL: MFCD01214137
CAS-Nummer: 349129-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR