Methyl-4-(trifluormethylthio)benzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-[(trifluoromethyl)sulphanyl]benzoate

CA-4712-5G CA-4712-1G
COBBCA-4712-5GEA 0 EUR
COBBCA-4712-5G COBBCA-4712-1G
Methyl-4-(trifluormethylthio)benzoat
Methyl-4-(trifluormethylthio)benzoat
Formel: C₉H₇F₃O₂S
MDL: MFCD06203725
CAS-Nummer: 88489-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR