4-Amino-N-(2,4-difluorphenyl)benzolsulfonamid

Lieferant: Thermo Scientific

RJF00762.250 RJF00762.1
MAYBRJF00762.250EA 0 EUR
MAYBRJF00762.250 MAYBRJF00762.1
4-Amino-N-(2,4-difluorphenyl)benzolsulfonamid
4-Amino-N-(2,4-difluorphenyl)benzolsulfonamid
Formel: C₁₂H₁₀F₂N₂O₂S
MDL: MFCD01935782
CAS-Nummer: 1717-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR