N-Butyl-4-fluorbenzolsulfonamid
Lieferant: COMBI-BLOCKS
OT-0960-1G
OT-0960-5G
COBBOT-0960-1GEA
0
EUR
COBBOT-0960-1G
COBBOT-0960-5G
N-Butyl-4-fluorbenzolsulfonamid
N-Butyl-4-fluorbenzolsulfonamid
Formel:
C₁₀H₁₄FNO₂S |
MDL:
MFCD01213077 CAS-Nummer: 312-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...