N-Ethyl-4-fluorbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0965-5G OT-0965-1G
COBBOT-0965-5GEA 0 EUR
COBBOT-0965-5G COBBOT-0965-1G
N-Ethyl-4-fluorbenzolsulfonamid
N-Ethyl-4-fluorbenzolsulfonamid
Formel: C₈H₁₀FNO₂S
MDL: MFCD01211991
CAS-Nummer: 383-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR