4-Fluor-N-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0779-5G OT-0779-1G
COBBOT-0779-5GEA 424 EUR
COBBOT-0779-5G COBBOT-0779-1G
4-Fluor-N-methylbenzolsulfonamid
4-Fluor-N-methylbenzolsulfonamid
Formel: C₇H₈FNO₂S
Molecular Weight: 189,2073 g/mol
MDL: MFCD01211989
CAS-Nummer: 433-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR