N-Cyclopropyl-4-fluorbenzolsulfonamid
Lieferant: COMBI-BLOCKS
OT-0973-1G
OT-0973-5G
COBBOT-0973-1GEA
0
EUR
COBBOT-0973-1G
COBBOT-0973-5G
N-Cyclopropyl-4-fluorbenzolsulfonamid
N-Cyclopropyl-4-fluorbenzolsulfonamid
Formel:
C₉H₁₀FNO₂S |
MDL:
MFCD02136916 CAS-Nummer: 425654-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...