4-Fluor-N-phenylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0962-5G OT-0962-1G
COBBOT-0962-5GEA 265 EUR
COBBOT-0962-5G COBBOT-0962-1G
4-Fluor-N-phenylbenzolsulfonamid
4-Fluor-N-phenylbenzolsulfonamid
Formel: C₁₂H₁₀FNO₂S
MDL: MFCD00447248
CAS-Nummer: 312-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR