Methyl-2-iod-4,5-dimethoxybenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 2-iodo-4,5-dimethoxybenzoate
HC-3355-1G
HC-3355-25G
HC-3355-5G
COBBHC-3355-1GEA
126
EUR
COBBHC-3355-1G
COBBHC-3355-25G
COBBHC-3355-5G
Methyl-2-iod-4,5-dimethoxybenzoat
Methyl-2-iod-4,5-dimethoxybenzoat
Formel:
C₁₀H₁₁IO₄ Molecular Weight: 322.0964 g/mol |
MDL:
MFCD09800786 CAS-Nummer: 173043-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...