3-Amino-N-tert-butylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: N-tert-Butyl 3-Aminophenylsulphonamide

OT-0132-5G OT-0132-1G
COBBOT-0132-5GEA 303 EUR
COBBOT-0132-5G COBBOT-0132-1G
3-Amino-N-tert-butylbenzolsulfonamid
3-Amino-N-tert-butylbenzolsulfonamid
Formel: C₁₀H₁₆N₂O₂S
Molecular Weight: 228,3100 g/mol
MDL: MFCD06660528
CAS-Nummer: 608523-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR