3-Amino-N-butylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0128-25G OT-0128-100G OT-0128-5G
COBBOT-0128-25GEA 257 EUR
COBBOT-0128-25G COBBOT-0128-100G COBBOT-0128-5G
3-Amino-N-butylbenzolsulfonamid
3-Amino-N-butylbenzolsulfonamid
Formel: C₁₀H₁₆N₂O₂S
MDL: MFCD00144861
CAS-Nummer: 143173-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR