N,N-Dimethyl-3-nitrobenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0117-5G OT-0117-1G
COBBOT-0117-5GEA 420 EUR
COBBOT-0117-5G COBBOT-0117-1G
N,N-Dimethyl-3-nitrobenzolsulfonamid
N,N-Dimethyl-3-nitrobenzolsulfonamid
Formel: C₈H₁₀N₂O₄S
MDL: MFCD01985504
CAS-Nummer: 26199-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR