N-tert-Butyl-3-nitrobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: N-(tert-Butyl)-3-nitrobenzenesulphonamide

OT-0115-1G OT-0115-5G
COBBOT-0115-1GEA 74.2 EUR
COBBOT-0115-1G COBBOT-0115-5G
N-tert-Butyl-3-nitrobenzolsulfonamid
N-tert-Butyl-3-nitrobenzolsulfonamid
Formel: C₁₀H₁₄N₂O₄S
Molecular Weight: 258,2942 g/mol
MDL: MFCD01985492
CAS-Nummer: 424818-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR