N-Butyl-3-nitrobenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0111-5G OT-0111-1G
COBBOT-0111-5GEA 204 EUR
COBBOT-0111-5G COBBOT-0111-1G
N-Butyl-3-nitrobenzolsulfonamid
N-Butyl-3-nitrobenzolsulfonamid
Formel: C₁₀H₁₄N₂O₄S
MDL: MFCD00144862
CAS-Nummer: 89840-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR