N-Cyclopropyl-3-nitrobenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0113-5G OT-0113-1G
COBBOT-0113-5GEA 303 EUR
COBBOT-0113-5G COBBOT-0113-1G
N-Cyclopropyl-3-nitrobenzolsulfonamid
N-Cyclopropyl-3-nitrobenzolsulfonamid
Formel: C₉H₁₀N₂O₄S
MDL: MFCD03015521
CAS-Nummer: 401589-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR