N-tert-Butylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0510-1G OT-0510-25G OT-0510-5G
COBBOT-0510-1GEA 98.3 EUR
COBBOT-0510-1G COBBOT-0510-25G COBBOT-0510-5G
N-tert-Butylbenzolsulfonamid
N-tert-Butylbenzolsulfonamid
Formel: C₁₀H₁₅NO₂S
Molecular Weight: 213,2966 g/mol
MDL: MFCD00462465
CAS-Nummer: 2512-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR