4-(3-Hydroxy-1-propinyl)benzonitril
Lieferant: Thermo Scientific
KM09036.250
KM09036.1
MAYBKM09036.250EA
0
EUR
MAYBKM09036.250
MAYBKM09036.1
4-(3-Hydroxy-1-propinyl)benzonitril
4-(3-Hydroxy-1-propinyl)benzonitril
Formel:
C₁₀H₇NO |
MDL:
MFCD00219780 CAS-Nummer: 80151-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...