3,5-Dinitro-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dinitro-2-methylbenzeneboronic acid

BB-2140-5G BB-2140-25G BB-2140-1G
COBBBB-2140-5GEA 98.3 EUR
COBBBB-2140-5G COBBBB-2140-25G COBBBB-2140-1G
3,5-Dinitro-2-methylphenylboronsäure
3,5-Dinitro-2-methylphenylboronsäure
Formel: C₇H₇BN₂O₆
Molecular Weight: 225,95 g/mol
MDL: MFCD00757434
CAS-Nummer: 24341-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR