4-Brom-N-isopropyl-3-methylbenzamid
Lieferant: COMBI-BLOCKS
Synonyme:
N-Isopropyl 4-bromo-3-methylbenzamide
CA-4555-5G
CA-4555-1G
COBBCA-4555-5GEA
303
EUR
COBBCA-4555-5G
COBBCA-4555-1G
4-Brom-N-isopropyl-3-methylbenzamid
4-Brom-N-isopropyl-3-methylbenzamid
Formel:
C₁₁H₁₄BrNO |
MDL:
MFCD09972122 CAS-Nummer: 1020252-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...