4-Brom-N-butyl-3-methylbenzamid

Lieferant: COMBI-BLOCKS

CA-4565-1G CA-4565-5G
COBBCA-4565-1GEA 149 EUR
COBBCA-4565-1G COBBCA-4565-5G
4-Brom-N-butyl-3-methylbenzamid
4-Brom-N-butyl-3-methylbenzamid
Formel: C₁₂H₁₆BrNO
MDL: MFCD11053785
CAS-Nummer: 1065073-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR