4-Brom-3-methyl-N-propylbenzamid

Lieferant: COMBI-BLOCKS

CA-4562-5G CA-4562-1G
COBBCA-4562-5GEA 451 EUR
COBBCA-4562-5G COBBCA-4562-1G
4-Brom-3-methyl-N-propylbenzamid
4-Brom-3-methyl-N-propylbenzamid
Formel: C₁₁H₁₄BrNO
MDL: MFCD09972127
CAS-Nummer: 1020252-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR