4-Brom-N-(2-hydroxyethyl)-3-methylbenzamid
Lieferant: COMBI-BLOCKS
CA-4556-5G
CA-4556-1G
COBBCA-4556-5GEA
884
EUR
COBBCA-4556-5G
COBBCA-4556-1G
4-Brom-N-(2-hydroxyethyl)-3-methylbenzamid
4-Brom-N-(2-hydroxyethyl)-3-methylbenzamid
Formel:
C₁₀H₁₂BrNO₂ |
MDL:
MFCD09972123 CAS-Nummer: 157672-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...