4-Brom-N-cyclohexyl-3-methylbenzamid
Lieferant: COMBI-BLOCKS
CA-4563-5G
CA-4563-1G
COBBCA-4563-5GEA
303
EUR
COBBCA-4563-5G
COBBCA-4563-1G
4-Brom-N-cyclohexyl-3-methylbenzamid
4-Brom-N-cyclohexyl-3-methylbenzamid
Formel:
C₁₄H₁₈BrNO |
MDL:
MFCD09972128 CAS-Nummer: 1020252-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...