4-Brom-3-methyl-N-phenylbenzamid

Lieferant: COMBI-BLOCKS

CA-4564-5G CA-4564-1G
COBBCA-4564-5GEA 303 EUR
COBBCA-4564-5G COBBCA-4564-1G
4-Brom-3-methyl-N-phenylbenzamid
4-Brom-3-methyl-N-phenylbenzamid
Formel: C₁₄H₁₂BrNO
MDL: MFCD09972129
CAS-Nummer: 1020252-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR