3-Bromtoluol-4-sulfonamid

Lieferant: COMBI-BLOCKS

OT-0172-5G OT-0172-1G
COBBOT-0172-5GEA 886 EUR
COBBOT-0172-5G COBBOT-0172-1G
3-Bromtoluol-4-sulfonamid
3-Bromtoluol-4-sulfonamid
Formel: C₇H₈BrNO₂S
MDL: MFCD11104196
CAS-Nummer: 89694-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR