3-Brom-N-butyl-4-methylbenzolsulfonamid
Lieferant: COMBI-BLOCKS
OT-0160-5G
OT-0160-1G
COBBOT-0160-5GEA
303
EUR
COBBOT-0160-5G
COBBOT-0160-1G
3-Brom-N-butyl-4-methylbenzolsulfonamid
3-Brom-N-butyl-4-methylbenzolsulfonamid
Formel:
C₁₁H₁₆BrNO₂S |
MDL:
MFCD07363833 CAS-Nummer: 850429-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...