3-Brom-N-methyl-5-methylbenzolsulfonamid
Lieferant: COMBI-BLOCKS
HC-6455-1G
HC-6455-5G
COBBHC-6455-1GEA
451
EUR
COBBHC-6455-1G
COBBHC-6455-5G
3-Brom-N-methyl-5-methylbenzolsulfonamid
3-Brom-N-methyl-5-methylbenzolsulfonamid
Formel:
C₈H₁₀BrNO₂S |
MDL:
MFCD09972147 CAS-Nummer: 1020252-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...