Methyl-3-chlor-4-methylbenzoat 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 3-chloro-4-methylbenzoate

H25933.14 H25933.06
H25933.14EA 282 EUR
H25933.14 H25933.06
Methyl-3-chlor-4-methylbenzoat 95%
Methyl-3-chlor-4-methylbenzoat

Purity: 95% CAS No.: 56525-63-4 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 27-29° Boiling Point: 119-120°/10mm MDL No.: MFCD00205141

Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Siedepunkt: 119…120 °C (10 mmHg)
Schmelzpunkt: 27…29 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00205141
CAS-Nummer: 56525-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR