Methyl-3-chlor-4-methylbenzoat

Lieferant: Thermo Scientific

Synonyme: Methyl 3-chloro-4-methylbenzoate

TL00703DA TL00703EA
MAYBTL00703DAEA 0 EUR
MAYBTL00703DA MAYBTL00703EA
Methyl-3-chlor-4-methylbenzoat
Methyl-3-chlor-4-methylbenzoat
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
MDL: MFCD00205141
CAS-Nummer: 56525-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR