3-Chlor-2-methylbenzolsulfonamid

Lieferant: FLUOROCHEM

CFF418.5 CFF418.10 CFF418.25 017440-5G 017440-10G 017440-25G 017440-1G
CHMPCFF418.5EA 348 EUR
CHMPCFF418.5 CHMPCFF418.10 CHMPCFF418.25 FLUO017440-5G FLUO017440-10G FLUO017440-25G FLUO017440-1G
3-Chlor-2-methylbenzolsulfonamid
3-Chlor-2-methylbenzolsulfonamid
Formel: C₇H₈ClNO₂S
Molecular Weight: 205,661 g/mol
MDL: MFCD00234295
CAS-Nummer: 3476-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR