Methyl-2-(2-methoxy-2-oxoethyl)benzoat

Lieferant: COMBI-BLOCKS

CA-4621-5G CA-4621-1G
COBBCA-4621-5GEA 692 EUR
COBBCA-4621-5G COBBCA-4621-1G
Methyl-2-(2-methoxy-2-oxoethyl)benzoat
Methyl-2-(2-methoxy-2-oxoethyl)benzoat
Formel: C₁₁H₁₂O₄
MDL: MFCD11053807
CAS-Nummer: 716-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR