3-Methoxycarbonyl-5-methylbenzoesäure

Lieferant: COMBI-BLOCKS

CA-4126-1G CA-4126-5G
COBBCA-4126-1GEA 114 EUR
COBBCA-4126-1G COBBCA-4126-5G
3-Methoxycarbonyl-5-methylbenzoesäure
3-Methoxycarbonyl-5-methylbenzoesäure
Formel: C₁₀H₁₀O₄
MDL: MFCD07363803
CAS-Nummer: 167299-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR