4-(Trifluormethyl)-N,N-dimethylbenzamid
Lieferant: COMBI-BLOCKS
Synonyme:
N,N-Dimethyl-4-(trifluoromethyl)benzamide
CA-4989-5G
CA-4989-1G
COBBCA-4989-5GEA
595
EUR
COBBCA-4989-5G
COBBCA-4989-1G
4-(Trifluormethyl)-N,N-dimethylbenzamid
4-(Trifluormethyl)-N,N-dimethylbenzamid
Formel:
C₁₀H₁₀F₃NO Molecular Weight: 217,1877 g/mol |
MDL:
MFCD16618942 CAS-Nummer: 25771-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...